Få mere at vide
2,3-Dihydro-5-benzofuranacetic Acid-d2, TRC
Artikelnummer. 30377875
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30377875 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377875 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4484875MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3-Dihydro-5-benzofuranacetic Acid-d2
CAS 1216878-16-8
Molekylær formel C10H8D2O3
Mængde 5 mg
Synonym (2,3-Dihydrobenzofuran-5-yl)acetic Acid-d2, 2,3-Dihydrobenzofuran-5-acetic Acid-d2, 2-(2,3-Dihydrobenzofuran-5-yl)acetic Acid-d2
InChI formel InChI=1S/C10H10O3/c11-10(12)6-7-1-2-9-8(5-7)3-4-13-9/h1-2,5H,3-4,6H2,(H,11,12)/i6D2
SMIL [2H]C([2H])(C(=O)O)c1ccc2OCCc2c1
IUPAC navn 2,2-dideuterio-2-(2,3-dihydro-1-benzofuran-5-yl)acetic acid
Molekylvægt (g/mol) 180.2
Formel vægt 180.0755