Learn More
2,3-Dihydro-1,4-benzodioxin-5-ylmethylamine, 97%, Thermo Scientific™
Catalog Number p-4509822
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
261633-71-0
C9H11NO2
165.192
OKRXRKUGZVMPKX-UHFFFAOYSA-N
2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine
2795040
2,3-dihydro-1,4-benzodioxin-5-ylmethanamine
C1COC2=C(C=CC=C2O1)CN
261633-71-0
C9H11NO2
165.192
OKRXRKUGZVMPKX-UHFFFAOYSA-N
2,3-dihydrobenzo b 1,4 dioxin-5-yl methanamine, c-2,3-dihydro-benzo 1,4 dioxin-5-yl-methylamine, 1,4-benzodioxin-5-methanamine, 2,3-dihydro, 2,3-dihydro-1,4-benzodioxin-5-ylmethylamine, 5-aminomethyl-1,4-benzodioxane, 1-2,3-dihydro-1,4-benzodioxin-5-yl methanamine, c-2,3-dihydrobenzo 1,4 dioxin-5-yl methylamine
2795040
2,3-dihydro-1,4-benzodioxin-5-ylmethanamine
C1COC2=C(C=CC=C2O1)CN
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.65
Percent Purity 95%
Chemical Name or Material 2, 3-Dihydro-1, 4-benzodioxin-5-ylmethylamine hydrochloride