Få mere at vide
2,3-Difluoropyridine, 97%, Thermo Scientific™
Artikelnummer. 11464340
Skift visning
Mængde:
250 mg
1 g
Pakningsstørrelse:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11464340 1 g 1g
11454340 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11464340 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H25771.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1513-66-2
Molekylær formel C5H3F2N
Molekylvægt (g/mol) 115.083
MDL nummer MFCD04114143
InChI nøgle OGVLEPMNNPZAPS-UHFFFAOYSA-N
Synonym 2,3-difluoro pyridine, difluoropyridine, pyridine, 2,3-difluoro, 2,3-difluorpyridine, 2,3-difluoropyridin, pubchem5414, 2,3-difiuoropyridine?, acmc-1bvua, 2,3-bis fluoranyl pyridine, ksc493a1t
PubChem CID 2783176
IUPAC navn 2,3-difluoropyridine
SMIL C1=CC(=C(N=C1)F)F
Tekniske data
Kemisk navn eller materiale 2,3-Difluoropyridine
Kogepunkt 125°C to 126°C
Flammepunkt 31°C (88°F)
Lugt Characteristic
Mængde 1 g
FN nummer UN1993
Formel vægt 115.08
Procent renhed 97%