missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Difluoro-6-methylpyridine, 98%

Brand:  Alfa Aesar™ H64514.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15471759

  • 2207.55 DKK / 5g


Estimated Shipment
03-08-2020
Add to basket

Specifications

Chemical Identifiers

1227579-04-5
129.11
HGBWAUANMSZOIJ-UHFFFAOYSA-N
70680696
CC1=NC(=C(C=C1)F)F
C6H5F2N
MFCD16610828
2,3-difluoro-6-methyl-pyridine
2,3-difluoro-6-methylpyridine

Specifications

2,3-Difluoro-6-methylpyridine
5g
C6H5F2N
2,3-difluoro-6-methyl-pyridine
CC1=NC(=C(C=C1)F)F
129.11
129.11
98%
Colorless to Yellow
1227579-04-5
MFCD16610828
HGBWAUANMSZOIJ-UHFFFAOYSA-N
2,3-difluoro-6-methylpyridine
70680696
Liquid
Videos
SDS Documents