missing translation for 'onlineSavingsMsg'
Learn More

2,3-Difluoro-6-methoxyphenol, 97%, Thermo Scientific™

Catalog Number 15473467
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
186306-70-7
C7H6F2O2
160.12
MFCD04116024
HDQMXTCQIQIJFG-UHFFFAOYSA-N
acmc-20ajxc, 2,3-difluoro 6-methoxy phenol, phenol, 2,3-difluoro-6-methoxy-9ci
3269728
2,3-difluoro-6-methoxyphenol
COC1=C(C(=C(C=C1)F)F)O
This item is not returnable. View return policy
186306-70-7
C7H6F2O2
160.12
MFCD04116024
HDQMXTCQIQIJFG-UHFFFAOYSA-N
acmc-20ajxc, 2,3-difluoro 6-methoxy phenol, phenol, 2,3-difluoro-6-methoxy-9ci
3269728
2,3-difluoro-6-methoxyphenol
COC1=C(C(=C(C=C1)F)F)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 44°C to 47°C
Quantity 250 mg
Formula Weight 160.12
Percent Purity 97%
Chemical Name or Material 2,3-Difluoro-6-methoxyphenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'