Learn More
2,3-Dichloropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific™
Catalog Number 11481740
Click to view available options
Quantity:
250 mg
1 g
1073353-78-2
C11H14BCl2NO2
273.95
MFCD06798257
LRQNNFMOVWWIHA-UHFFFAOYSA-N
2,3-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,3-dichloropyridine-4-boronic acid pinacol ester, pyridine, 2,3-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-13-5531, 2,3-dichloropyridine-4-boronic acid, pinacol ester, 2,3-dichloropyridin-4-yl boronic acid pinacol ester, 2,3-dichloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
44755158
2,3-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=C(Cl)C(Cl)=NC=C1
1073353-78-2
C11H14BCl2NO2
273.95
MFCD06798257
LRQNNFMOVWWIHA-UHFFFAOYSA-N
2,3-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,3-dichloropyridine-4-boronic acid pinacol ester, pyridine, 2,3-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-13-5531, 2,3-dichloropyridine-4-boronic acid, pinacol ester, 2,3-dichloropyridin-4-yl boronic acid pinacol ester, 2,3-dichloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
44755158
2,3-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=C(Cl)C(Cl)=NC=C1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 83°C to 85°C
Quantity 1 g
Formula Weight 273.96
Percent Purity 95%
Chemical Name or Material 2,3-Dichloropyridine-4-boronic acid pinacol ester