missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-6-fluorobenzonitrile, 97%, Thermo Scientific™

Catalog Number 11486040
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
79544-26-6
C7H2Cl2FN
190.00
MFCD04115878
PISOOQLALMAJRE-UHFFFAOYSA-N
2,3-dichloro-6-fluorobenzenecarbonitrile
3767163
2,3-dichloro-6-fluorobenzonitrile
FC1=C(C#N)C(Cl)=C(Cl)C=C1
This item is not returnable. View return policy
79544-26-6
C7H2Cl2FN
190.00
MFCD04115878
PISOOQLALMAJRE-UHFFFAOYSA-N
2,3-dichloro-6-fluorobenzenecarbonitrile
3767163
2,3-dichloro-6-fluorobenzonitrile
FC1=C(C#N)C(Cl)=C(Cl)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3439
Formula Weight 190
Percent Purity 97%
Chemical Name or Material 2,3-Dichloro-6-fluorobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'