missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3-Dichloro-6-fluorobenzonitrile, 97%

Brand:  Alfa Aesar™ H26584.06

Additional Details : CAS Number : 79544-26-6 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11496040

  • POA / 5g

Currently not in stock

Please to purchase this item. Need a web account? Register with us today!

This item cannot be ordered online because the price is subject to change. Please contact our Product Support team on +45 70 27 99 20 for assistance.
View alternative products »


Description

Description

Specifications

Chemical Identifiers

79544-26-6
189.998
PISOOQLALMAJRE-UHFFFAOYSA-N
3767163
C1=CC(=C(C(=C1F)C#N)Cl)Cl
C7H2Cl2FN
MFCD04115878
2,3-dichloro-6-fluorobenzenecarbonitrile
2,3-dichloro-6-fluorobenzonitrile

Specifications

2,3-Dichloro-6-fluorobenzonitrile
79544-26-6
MFCD04115878
2,3-dichloro-6-fluorobenzenecarbonitrile
C1=CC(=C(C(=C1F)C#N)Cl)Cl
189.998
190
5g
C7H2Cl2FN
UN3439
PISOOQLALMAJRE-UHFFFAOYSA-N
2,3-dichloro-6-fluorobenzonitrile
3767163
97%
Videos
SDS Documents