missing translation for 'onlineSavingsMsg'
Få mere at vide

2,3-Dibromo-1-propan-1,1,2,3,3-d5-ol, TRC

Artikelnummer. 30374761
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30374761 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30374761 Leverandør Toronto Research Chemicals Leverandørnr. TRCD42552210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2,3-Dibromo-1-propan-1,1,2,3,3-d5-ol
CAS 117662-57-4
Molekylær formel C3HD5Br2O
Mængde 10 mg
Synonym 2,3-Dibromo-1-propanol-d5, 2,3-Dibromopropyl Acohol-d5, Brominex 257-d5, DBP-d5, DBP (flame retardant)-d5, NSC 6203-d5, 1,2-Dibromopropan-3-ol-d5
InChI formel InChI=1S/C3H6Br2O/c4-1-3(5)2-6/h3,6H,1-2H2/i1D2,2D2,3D
SMIL BrC([2H])([2H])C(Br)([2H])C([2H])([2H])O
Molekylvægt (g/mol) 222.92
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.