Få mere at vide
2-(3-Chlorophenyl)-1-(3-pyridinyl-d4)-1-ethanone, TRC
Artikelnummer. 30367202
Skift visning
Mængde:
20 mg
Artikelnummer. Mængde
30367202 20 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367202 Leverandør Toronto Research Chemicals Leverandørnr. TRCC37960720MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(3-Chlorophenyl)-1-(3-pyridinyl-d4)-1-ethanone
CAS 1189880-12-3
Molekylær formel C13H6D4ClNO
Mængde 20 mg
Synonym m-Chlorobenzyl 3-Pyridyl Ketone-d4
InChI formel InChI=1S/C13H10ClNO/c14-12-5-1-3-10(7-12)8-13(16)11-4-2-6-15-9-11/h1-7,9H,8H2/i2D,4D,6D,9D
SMIL [2H]c1nc([2H])c(C(=O)Cc2cccc(Cl)c2)c([2H])c1[2H]
IUPAC navn 2-(3-chlorophenyl)-1-(2,4,5,6-tetradeuteriopyridin-3-yl)ethanone
Molekylvægt (g/mol) 235.7
Formel vægt 235.0702