Få mere at vide
2,3-Butanediol-d8 (Mixture of Diastereomers), TRC
Artikelnummer. 30337656
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30337656 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30337656 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69002950MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3-Butanediol-d8
CAS 347841-77-4
Molekylær formel C4 D8 H2 O2
Mængde 50 mg
Synonym 2,3-Butane-1,1,1,2,3,4,4,4-d8-diol (9CI), 2,3-Butanediol-d8, 2,3-Butanediol-D8, 2,3-Butanediol-1,1,1,2,3,4,4,4-d8, 2,3-Butane-d8-diol
InChI formel InChI=1S/C4H10O2/c1-3(5)4(2)6/h3-6H,1-2H3/i1D3,2D3,3D,4D
SMIL [2H]C([2H])([2H])C([2H])(O)C([2H])(O)C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,3,4,4,4-octadeuteriobutane-2,3-diol
Molekylvægt (g/mol) 98.17
Formel vægt 98.118