missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Bromomethyl)phenyl]-4,5-diphenylthiazole, 97%, Thermo Scientific™

Catalog Number p-7034142
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1217863-18-7
C22H16BrNS
406.34
MFCD16251571
ZIAGAOYXDFEXQY-UHFFFAOYSA-N
2-3-bromomethyl phenyl-4,5-diphenylthiazole, 2-3-bromomethyl phenyl-4,5-diphenyl-1,3-thiazole
73012488
2-[3-(bromomethyl)phenyl]-4,5-diphenyl-1,3-thiazole
BrCC1=CC=CC(=C1)C1=NC(=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1217863-18-7
C22H16BrNS
406.34
MFCD16251571
ZIAGAOYXDFEXQY-UHFFFAOYSA-N
2-3-bromomethyl phenyl-4,5-diphenylthiazole, 2-3-bromomethyl phenyl-4,5-diphenyl-1,3-thiazole
73012488
2-[3-(bromomethyl)phenyl]-4,5-diphenyl-1,3-thiazole
BrCC1=CC=CC(=C1)C1=NC(=C(S1)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 406.34
Percent Purity 97%
Chemical Name or Material 2-[3-(Bromomethyl)phenyl]-4,5-diphenylthiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'