Learn More
Alfa Aesar™ 2-(3-Benzoylphenyl)-N-tert-butylpropionamide, 95%
Brand:  Alfa Aesar™ H65051.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15402459
  • 2009.39 DKK / 1g
Estimated Shipment date
from Supplier 19-11-2018
Add to basket
Specifications
Chemical Identifiers
MFCD28098742
Specifications
2-(3-Benzoylphenyl)-N-tert-butylpropionamide
MFCD28098742
Solid
1g
309.4
95%