missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(4-bromo-2-fluorophenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65800.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469906

  • 1908.81 DKK / 1g


Estimated Shipment date
from Supplier 17-01-2019
Add to basket

Specifications

Chemical Identifiers

MFCD28098826

Specifications

2-(3-Benzoylphenyl)-N-(4-bromo-2-fluorophenyl)propionamide
MFCD28098826
Solid
1g
426.28
95%
Videos
SDS