missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Benzoylphenyl)-N-(3-chloro-4-fluorophenyl)propionamide, 95%

Brand:  Alfa Aesar™ H65398.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469616

  • 1908.81 DKK / 1g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

Specifications

2-(3-Benzoylphenyl)-N-(3-chloro-4-fluorophenyl)propionamide
1g
Solid
White
381.83
95%
Videos
SDS