missing translation for 'onlineSavingsMsg'
Få mere at vide

2-(3-Amino-4-chlorobenzoyl)benzoic Acid), TRC

Artikelnummer. 30328032
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 g
Pakningsstørrelse:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30328032 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30328032 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6032002.5G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-(3-Amino-4-chlorobenzoyl)benzoic acid
CAS 118-04-7
Molekylær formel C14H10ClNO3
Mængde 2.5 g
Synonym o-(3-Amino-4-chlorobenzoyl)-benzoic acid, 2-[(3-Amino-4-chlorophenyl)carbonyl]benzoic acid, 3'-Amino-4'-chloro-o-benzoylbenzoic acid, 4-Chloro-3-aminobenzophenone-2'-carboxylic acid, NSC 74496, o-(3-Amino-4-chlorobenzoyl)benzoic acid
InChI formel InChI=1S/C14H10ClNO3/c15-11-6-5-8(7-12(11)16)13(17)9-3-1-2-4-10(9)14(18)19/h1-7H,16H2,(H,18,19)
SMIL O=C(C1=CC(N)=C(Cl)C=C1)C(C=CC=C2)=C2C(O)=O
IUPAC navn 2-(3-amino-4-chlorobenzoyl)benzoic acid
Molekylvægt (g/mol) 275.69
Formel vægt 275.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.