Få mere at vide
2,3,6-Trichloroanisole-d3 (methoxy-d3), CDN
Artikelnummer. 30318634
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318634 0.1 g
30309380 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318634 Leverandør CDN Leverandørnr. CDND59510.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,6-Trichloroanisole D3 (methyl D3)
CAS 1219798-64-7
Molekylær formel C7 D3 H2 Cl3 O
Mængde 0.1 g
Synonym 2,3,6-Trichloroanisole D3 (methyl D3), 2,3,6-Trichloroanisole-d3, 1,2,4-Trichloro-3-(methoxy-d3)benzene
InChI formel InChI=1 S/C7H5Cl3O/c1-11-7-5(9)3-2-4(8)6(7)10/h2-3 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])Oc1c(Cl)ccc(Cl)c1Cl
IUPAC navn 1,2,4-trichloro-3-(trideuteriomethoxy)benzene
Molekylvægt (g/mol) 214.4915
Formel vægt 212.9594 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity