missing translation for 'onlineSavingsMsg'
Få mere at vide

2,3,5-Trimethylpyridine N-Oxide, TRC

Artikelnummer. 30456508
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456508 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30456508 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79670510G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2,3,5-Trimethylpyridine N-Oxide
CAS 74409-42-0
Molekylær formel C8 H11 N O
Mængde 10 g
Synonym Pyridine','2,3,5-trimethyl-','1-oxide, 2,3,5-Collidine N-oxide, 2,3,5-Trimethyl-1-oxidopyridin-1-ium, 2,3,5-Trimethylpyridine 1-oxide, 2,3,5-Trimethylpyridine N-oxide
InChI formel InChI=1S/C8H11NO/c1-6-4-7(2)8(3)9(10)5-6/h4-5H,1-3H3
SMIL Cc1cc(C)c(C)[n+]([O-])c1
IUPAC navn 2,3,5-trimethyl-1-oxidopyridin-1-ium
Molekylvægt (g/mol) 137.18
Formel vægt 137.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.