missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,5-Trimethylpyrazine, 99%

Brand:  Alfa Aesar™ A14870.18

Additional Details : CAS Number : 14667-55-1 Weight : 0.05000kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 50g

 View more versions of this product

Product Code. 11492657

  • 703.00 DKK / 50g

Estimated Shipment
28-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

14667-55-1
122.171
IAEGWXHKWJGQAZ-UHFFFAOYSA-N
26808
CC1=CN=C(C(=N1)C)C
C7H10N2
MFCD00006145
trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
2,3,5-trimethylpyrazine

Specifications

2,3,5-Trimethylpyrazine
54°C (129°F)
171°C to 172°C
14667-55-1
MFCD00006145
2423
IAEGWXHKWJGQAZ-UHFFFAOYSA-N
2,3,5-trimethylpyrazine
26808
99%
0.979
1.505
50g
C7H10N2
UN1993
trimethylpyrazine, pyrazine, trimethyl, 2,3,5-trimethyl pyrazine, 2,3,6-trimethylpyrazine, ccris 2932, unii-q8pr0w8tit, trimethyl pyrazine, trimethyl-pyrazine, fema no. 3244, pyrazine, 2,3,5-trimethyl
CC1=CN=C(C(=N1)C)C
122.171
122.17
Videos
SDS Documents