Få mere at vide
2,3,5-Trimethylphenol-d11, TRC
Artikelnummer. 30179636
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179636 10 mg
30178971 50 mg
30179013 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179636 Leverandør Toronto Research Chemicals Leverandørnr. TRCT79659610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 362049-46-5
Molekylær formel C9 D11 H O
Molekylvægt (g/mol) 147.2588
Synonym Phen-2,4-d2-ol, 3,5,6-tri(methyl-d3)- (9CI), 2,3,5-Trimethylphenol-d11, 2,3,5-Trimethylphenol-D11, 2,3,5-Tri(methyl-d3)phenol-4,6-d2, 3,5,6-Tri(methyl-d3)phenol-2,4-d2
IUPAC navn 2,4-dideuterio-3,5,6-tris(trideuteriomethyl)phenol
SMIL [2H]c1c(O)c(c(c([2H])c1C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale 2,3,5-Trimethylphenol-d11
Mængde 10 mg
InChI formel InChI=1S/C9H12O/c1-6-4-7(2)8(3)9(10)5-6/h4-5,10H,1-3H3/i1D3,2D3,3D3,4D,5D
Formel vægt 147.1579