missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,5-Trichloropyridine, ≥98%

Brand:  Alfa Aesar™ H64173.A1

Additional Details : CAS Number : 16063-70-0 Weight : 1.00000kg

Quantity 1kg

 View more versions of this product

Product Code. 15471259

  • 3236.75 DKK / 1kg


Estimated Shipment date
from Supplier 30-06-2020
Add to basket

Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Chemical Identifiers

16063-70-0
182.428
CNLIIAKAAMFCJG-UHFFFAOYSA-N
27666
C1=C(C=NC(=C1Cl)Cl)Cl
C5H2Cl3N
MFCD00043007
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
2,3,5-trichloropyridine

Specifications

2,3,5-Trichloropyridine
219°C (literature)
46°C to 50°C (literature)
16063-70-0
MFCD00043007
pyridine, 2,3,5-trichloro, 2,3,5-trichloro pyridine, 2,3,5-trichloro-pyridine, pubchem1127, g00013-watson-int, acmc-1at5w, 2,3,6-trichloro-pyridine, 2, 3, 5-trichloropyridine, ksc179q6d, tpc-i005
CNLIIAKAAMFCJG-UHFFFAOYSA-N
2,3,5-trichloropyridine
27666
Solid
113°C (235°F)
White to Yellow
1kg
C5H2Cl3N
119384
Slightly soluble in water.
C1=C(C=NC(=C1Cl)Cl)Cl
182.428
182.43
≥98%
Videos
SDS Documents