Få mere at vide
2,3,5-Trichlorobenzoic Acid, TRC
Artikelnummer. 30451897
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30451897 1 g
30454869 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30451897 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7743451G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 50-73-7
Molekylær formel C7 H3 Cl3 O2
Molekylvægt (g/mol) 225.46
Synonym Benzoic acid, 2,3,5-trichloro-, 2,3,5-Trichlorobenzoic acid
IUPAC navn 2,3,5-trichlorobenzoic acid
SMIL OC(=O)c1cc(Cl)cc(Cl)c1Cl
Tekniske data
Kemisk navn eller materiale 2,3,5-Trichlorobenzoic acid
Mængde 1 g
InChI formel InChI=1S/C7H3Cl3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,(H,11,12)
Formel vægt 223.9199