missing translation for 'onlineSavingsMsg'
Få mere at vide

2,3,5,6-Tetraphenylpyrazine, TRC

Produktkod. 30420632
Change view
Klicka för att se tillgängliga alternativ
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Mængde
30420632 500 mg
30411297 250 mg
30418496 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30420632 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT204561500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 642-04-6
Molekylær formel C28H20N2
Molekylvægt (g/mol) 384.47
Synonym 2,3,5,6-Tetraphenyl-1,4-pyrazine, Amaron, NSC 123074, Tetraphenylpyrazine
IUPAC navn 2,3,5,6-tetraphenylpyrazine
SMIL c1ccc(cc1)c2nc(c3ccccc3)c(nc2c4ccccc4)c5ccccc5

Specifications

Kemisk navn eller materiale 2,3,5,6-Tetraphenylpyrazine
Mængde 500 mg
InChI formel InChI=1S/C28H20N2/c1-5-13-21(14-6-1)25-26(22-15-7-2-8-16-22)30-28(24-19-11-4-12-20-24)27(29-25)23-17-9-3-10-18-23/h1-20H
Formel vægt 384.1626
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.