missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorothiophenol, 98%, Thermo Scientific Chemicals

Catalog Number p-7027038
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
769-40-4
C6H2F4S
182.136
MFCD00004829
IGOGJHYWSOZGAE-UHFFFAOYSA-N
2,3,5,6-tetrafluorothiophenol, 2,3,5,6-tetrafluorobenzene-1-thiol, benzenethiol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorbenzolthiol, timtec-bb sbb000230, #, 2,3,5,6-tetrafluorophenyl hydrosulfide, 2,3,5,6-tetrafluorobenzenethiol
69859
2,3,5,6-tetrafluorobenzenethiol
C1=C(C(=C(C(=C1F)F)S)F)F
This item is not returnable. View return policy
769-40-4
C6H2F4S
182.136
MFCD00004829
IGOGJHYWSOZGAE-UHFFFAOYSA-N
2,3,5,6-tetrafluorothiophenol, 2,3,5,6-tetrafluorobenzene-1-thiol, benzenethiol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorbenzolthiol, timtec-bb sbb000230, #, 2,3,5,6-tetrafluorophenyl hydrosulfide, 2,3,5,6-tetrafluorobenzenethiol
69859
2,3,5,6-tetrafluorobenzenethiol
C1=C(C(=C(C(=C1F)F)S)F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.53
Boiling Point 152°C to 153°C
Flash Point 48°C (119°F)
Odor Stench
Refractive Index 1.485
Quantity 5 g
UN Number UN1992
Sensitivity Air Sensitive
Formula Weight 182.14
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluorothiophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'