missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,5,6-Tetrafluoropyridine, 97%

Brand:  Alfa Aesar™ B21905.06

Additional Details : CAS Number : 2875-18-5 Weight : 0.00500kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11494977

  • 1346.00 DKK / 5g

Estimated Shipment
18-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

2875-18-5
151.064
HWIPMBCMGVXOKN-UHFFFAOYSA-N
137749
C1=C(C(=NC(=C1F)F)F)F
C5HF4N
MFCD00011734
pyridine, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-pyridine, 2,3,5,6-tetrafluoropyridine, pubchem9303, acmc-209h4j, ksc494q4b, hwipmbcmgvxokn-uhfffaoysa, 2,3,5,6-tetrafluoro-pyridine; pyridine, 2,3,5,6-tetrafluoro
2,3,5,6-tetrafluoropyridine

Specifications

2,3,5,6-Tetrafluoropyridine
30°C (86°F)
102°C
2875-18-5
MFCD00011734
pyridine, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-pyridine, 2,3,5,6-tetrafluoropyridine, pubchem9303, acmc-209h4j, ksc494q4b, hwipmbcmgvxokn-uhfffaoysa, 2,3,5,6-tetrafluoro-pyridine; pyridine, 2,3,5,6-tetrafluoro
C1=C(C(=NC(=C1F)F)F)F
151.064
151.06
1.499
1.4046
5g
C5HF4N
UN1993
HWIPMBCMGVXOKN-UHFFFAOYSA-N
2,3,5,6-tetrafluoropyridine
137749
97%
Videos
SDS Documents