Få mere at vide
2,3,5,6-Tetrachloroaniline-d3, CDN
Artikelnummer. 30319002
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319002 0.25 g -
30316410 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319002 Leverandør CDN Leverandørnr. CDND69110.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,5,6-Tetrachloroaniline-D3
CAS 1219806-05-9
Molekylær formel C6 D3 Cl4 N
Mængde 0.25 g
Synonym 2,3,5,6-Tetrachlorobenzene-4-d-amine-d2, 2,3,5,6-Tetrachloroaniline-d3, 2,3,5,6-Tetrachloroaniline-D3
InChI formel InChI=1 S/C6H3Cl4N/c7-2-1-3(8)5(10)6(11)4(2)9/h1H,11H2/i1D/hD2
SMIL [2 H]N([2 H])c1c(Cl)c(Cl)c([2 H])c(Cl)c1Cl
IUPAC navn 2,3,5,6-tetrachloro-N,N,4-trideuterioaniline
Molekylvægt (g/mol) 233.9252
Formel vægt 231.9208 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity