Få mere at vide
2,3,5,6-Tetrachloro-p-xylene, TRC
Artikelnummer. 30414630
Skift visning
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30414630 1 g
30411666 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30414630 Leverandør Toronto Research Chemicals Leverandørnr. TRCT2913501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 877-10-1
Molekylær formel C8H6Cl4
Molekylvægt (g/mol) 243.95
Synonym 1,2,4,5-Tetrachloro-3,6-dimethylbenzene, 1,2,4,5-Tetrachlorodimethylbenzene, 1,2,4,5-Tetrachloroxylene, NSC 66546, Tetrachloro-p-xylene
IUPAC navn 1,2,4,5-tetrachloro-3,6-dimethylbenzene
SMIL Cc1c(Cl)c(Cl)c(C)c(Cl)c1Cl
Tekniske data
Kemisk navn eller materiale 2,3,5,6-Tetrachloro-p-xylene
Mængde 1 g
InChI formel InChI=1S/C8H6Cl4/c1-3-5(9)7(11)4(2)8(12)6(3)10/h1-2H3
Formel vægt 241.9224