Få mere at vide
2,3,4,-Tri-O-benzyl-L-fucopyranose, TRC
Artikelnummer. 30456737
Skift visning
Mængde:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30456737 1 g
30455865 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30456737 Leverandør Toronto Research Chemicals Leverandørnr. TRCT7680001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,4,-Tri-O-benzyl-L-fucopyranose
CAS 60431-34-7
Molekylær formel C27 H30 O5
Mængde 1 g
InChI formel InChI=1S/C27H30O5/c1-20-24(29-17-21-11-5-2-6-12-21)25(30-18-22-13-7-3-8-14-22)26(27(28)32-20)31-19-23-15-9-4-10-16-23/h2-16,20,24-28H,17-19H2,1H3/t20-,24+,25+,26-,27?/m0/s1
SMIL C[C@@H]1OC(O)[C@@H](OCc2ccccc2)[C@H](OCc3ccccc3)[C@@H]1OCc4ccccc4
IUPAC navn (3S,4R,5R,6S)-6-methyl-3,4,5-tris(phenylmethoxy)oxan-2-ol
Molekylvægt (g/mol) 434.52
Formel vægt 434.2093