Få mere at vide
2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride, Mikromol™
Artikelnummer. 30051573
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051573 100 mg
30051001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051573 Leverandør Mikromol Leverandørnr. MM0022.08
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride
CAS 13078-76-7
Analyt- eller komponentnavne 2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C11 H17 N O2 . Cl H
Certificeringer/compliance ISO 17025
Synonym Verapamil Hydrochloride Imp. B (EP), Verapamil Imp. B (EP), 2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride, Verapamil Hydrochloride Imp. B (EP) as Hydrochloride, Verapamil Hydrochloride Impurity B as Hydrochloride, Verapamil Impurity B as Hydrochloride
InChI formel InChI=1S/C11H17NO2.ClH/c1-12-7-6-9-4-5-10(13-2)11(8-9)14-3;/h4-5,8,12H,6-7H2,1-3H3;1H
SMIL Cl.CNCCc1ccc(OC)c(OC)c1
IUPAC navn 2-(3,4-dimethoxyphenyl)-N-methylethanamine;hydrochloride
Molekylvægt (g/mol) 231.72
Mængde 100 mg
Formel vægt 231.1026 g/mol