Få mere at vide
2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride, Mikromol™
Beskrivelse
2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride |
| CAS | 13078-76-7 |
| Analyt- eller komponentnavne | 2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H17 N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Verapamil Hydrochloride Imp. B (EP), Verapamil Imp. B (EP), 2-(3,4-Dimethoxyphenyl)-N-methylethanamine Hydrochloride, Verapamil Hydrochloride Imp. B (EP) as Hydrochloride, Verapamil Hydrochloride Impurity B as Hydrochloride, Verapamil Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C11H17NO2.ClH/c1-12-7-6-9-4-5-10(13-2)11(8-9)14-3;/h4-5,8,12H,6-7H2,1-3H3;1H |
| SMIL | Cl.CNCCc1ccc(OC)c(OC)c1 |
| IUPAC navn | 2-(3,4-dimethoxyphenyl)-N-methylethanamine;hydrochloride |
| Vis mere |