Få mere at vide
2-(3,4-Dimethoxy-d6-phenyl)ethyl-1,1,2,2-d4-amine HCl, CDN
Artikelnummer. 30310578
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310578 0.01 g
30325102 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310578 Leverandør CDN Leverandørnr. CDND74230.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(3,4-Dimethoxyphenyl)ethanamine-D10 (aminoethyl-D4,dimethoxy-D6) Hydrochloride
CAS 1398065-73-0
Molekylær formel C10 D10 H5 N O2 . Cl H
Mængde 0.01 g
Synonym 3,4-Di(methoxy-d3)benzeneethan-α,α,β,β-d4-amine hydrochloride (1:1), α,α,β,β-Tetradeuterio-3,4-bis(trideuteriomethoxy)phenylethylamine hydrochloride, 3,4-Di-O-methyldopamine-d10 hydrochloride, 3,4-Di(methoxy-d3)-β-phenylethylamine-α,α,β,β-d4 hydrochloride, Homoveratrylamine-d10 hydrochloride, 2-(3,4-Dimethoxyphenyl)ethanamine-D10 (aminoethyl-D4,dimethoxy-D6) hydrochloride, 2-(3,4-Dimethoxy-d6-phenyl)ethyl-1,1,2,2-d4-amine HCl
InChI formel InChI=1 S/C10H15NO2.ClH/c1-12-9-4-3-8(5-6-11)7-10(9)13-2;/h3-4,7 H,5-6,11H2,1-2H3;1 H/i1D3,2D3,5D2,6D2;
SMIL Cl.[2 H]C([2 H])([2 H])Oc1ccc(cc1OC([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])N
IUPAC navn 2-[3,4-bis(trideuteriomethoxy)phenyl]-1,1,2,2-tetradeuterioethanamine;hydrochloride
Molekylvægt (g/mol) 227.7542
Formel vægt 227.1497 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity