Få mere at vide
2,3,4,7,8-Pentachlorodibenzofuran (~90%), TRC
Artikelnummer. 30136954
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136954 50 mg
30010755 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136954 Leverandør Toronto Research Chemicals Leverandørnr. TRCP23797550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 57117-31-4
Molekylær formel C12 H3 Cl5 O
Molekylvægt (g/mol) 340.42
Synonym Dibenzofuran, 2,3,4,7,8-pentachloro-, 2,3,4,7,8-Pentachlorodibenzofuran, 2,3,4,7,8-PCDF, 2,3,4,7,8-PeCDF, 23478-PCDF, F 114, PCDF, PCDF 114, PECDF, 2,3,4,7,8-PentaCDF, 23478-PentaCDF, 23478-Pentachlorodibenzofuran
IUPAC navn 2,3,4,7,8-pentachlorodibenzofuran
SMIL Clc1cc2oc3c(Cl)c(Cl)c(Cl)cc3c2cc1Cl
Tekniske data
Kemisk navn eller materiale 2,3,4,7,8-Pentachlorodibenzofuran
Mængde 50 mg
InChI formel InChI=1S/C12H3Cl5O/c13-6-1-4-5-2-8(15)10(16)11(17)12(5)18-9(4)3-7(6)14/h1-3H
Formel vægt 337.8627