missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%, Thermo Scientific Chemicals

Catalog Number p-7059877
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
53081-25-7
C34H36O6
MFCD00132927
This item is not returnable. View return policy
53081-25-7
C34H36O6
MFCD00132927

2,3,4,6-Tetra-O-benzyl-D-galactopyranose is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 540.65
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 2,3,4,6-Tetra-O-benzyl-D-galactopyranose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'