Learn More
2,3,4,6-Tetra-O-acetyl-D-glucopyranose, TRC
Product Code. 30411852
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411852 2.5 g 2.5g
30413222 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30411852 Supplier Toronto Research Chemicals Supplier No. TRCT2800002.5G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10343-06-3
Molecular Formula C14 H20 O10
Molecular Weight (g/mol) 348.3
Synonym D-Glucopyranose, 2,3,4,6-tetraacetate, Glucopyranose, 2,3,4,6-tetraacetate, D- (8CI), 2,3,4,6-Tetra-O-acetyl-D-glucopyranose, 2,3,4,6-Tetra-O-acetyl-D-glucose
IUPAC Name [(2R,3R,4S,5R)-3,4,5-triacetyloxy-6-hydroxyoxan-2-yl]methyl acetate
SMILES CC(=O)OC[C@H]1OC(O)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C14H20O10/c1-6(15)20-5-10-11(21-7(2)16)12(22-8(3)17)13(14(19)24-10)23-9(4)18/h10-14,19H,5H2,1-4H3/t10-,11-,12+,13-,14?/m1/s1
Formula Weight 348.1056
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-D-glucopyranose