Få mere at vide
2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranosylamine, TRC
Artikelnummer. 30409387
Skift visning
Mængde:
100 mg
250 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30409387 100 mg -
30414771 2.5 g 2.5g
30419046 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30409387 Leverandør Toronto Research Chemicals Leverandørnr. TRCT279550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58484-22-3
Molekylær formel C14H21NO9
Molekylvægt (g/mol) 347.32
Synonym β-D-Galactopyranosylamine 2,3,4,6-Tetraacetate
IUPAC navn [(2R,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-aminooxan-2-yl]methyl acetate
SMIL CC(=O)OC[C@H]1O[C@@H](N)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C
Tekniske data
Kemisk navn eller materiale 2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranosylamine
Mængde 100 mg
InChI formel InChI=1S/C14H21NO9/c1-6(16)20-5-10-11(21-7(2)17)12(22-8(3)18)13(14(15)24-10)23-9(4)19/h10-14H,5,15H2,1-4H3/t10-,11+,12+,13-,14-/m1/s1
Formel vægt 347.1216