missing translation for 'onlineSavingsMsg'
Learn More

2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%, Thermo Scientific Chemicals

Catalog Number 15490517
Click to view available options
Quantity:
500 mg
1 g
5 g
This item is not returnable. View return policy
17682-70-1
C12H20O6
260.286
MFCD00132929
GQXSDDHYUVYJCQ-HNBLOZHYSA-N
92220487
[(3aS,4aR,8aR,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxin-3a-yl]methanol
CC1(OCC2C(O1)C3C(O2)(OC(O3)(C)C)CO)C
This item is not returnable. View return policy
17682-70-1
C12H20O6
260.286
MFCD00132929
GQXSDDHYUVYJCQ-HNBLOZHYSA-N
92220487
[(3aS,4aR,8aR,8bR)-2,2,7,7-tetramethyl-4a,5,8a,8b-tetrahydro-[1,3]dioxolo[3,4]furo[1,3-d][1,3]dioxin-3a-yl]methanol
CC1(OCC2C(O1)C3C(O2)(OC(O3)(C)C)CO)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 80°C
Color White
Quantity 5 g
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'