missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number p-7023785
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
832-53-1
C6ClF5O2S
266.566
MFCD00007427
UOJCTEGNHXRPKO-UHFFFAOYSA-N
pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
70026
2,3,4,5,6-pentafluorobenzenesulfonyl chloride
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F
This item is not returnable. View return policy
832-53-1
C6ClF5O2S
266.566
MFCD00007427
UOJCTEGNHXRPKO-UHFFFAOYSA-N
pentafluorobenzenesulfonyl chloride, 2,3,4,5,6-pentafluorobenzene-1-sulfonyl chloride, pentafluorobenzenesulphonyl chloride, benzenesulfonyl chloride, pentafluoro, pentafluorophenylsulfonyl chloride, 2,3,4,5,6-pentafluorobenzenesulfonyl choride, 2,3,4,5,6-pentafluoro-benzenesulfonyl chloride, chloro 2,3,4,5,6-pentafluorophenyl sulfone, pubchem5683, pentafluorobenzenesulfonylchloride
70026
2,3,4,5,6-pentafluorobenzenesulfonyl chloride
C1(=C(C(=C(C(=C1F)F)S(=O)(=O)Cl)F)F)F

Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

Solubility
Soluble in water (reacts slowly).

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases, water/moisture. Keep the container tightly closed in a cool, dry place. Store under inert gas.
TRUSTED_SUSTAINABILITY
Density 1.796
Boiling Point 210°C to 211°C
Flash Point 90°C (194°F)
Refractive Index 1.479
Quantity 1 g
UN Number UN3265
Beilstein 2142325
Sensitivity Moisture sensitive
Solubility Information Soluble in water (reacts slowly).
Formula Weight 266.57
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'