Få mere at vide
2,3,4,5,6-Pentachlorobiphenyl-2',3',4',5',6'-d5, CDN
Artikelnummer. 30314992
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30314992 0.005 g
30310163 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314992 Leverandør CDN Leverandørnr. CDND64930.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,4,5,6-Pentachlorobiphenyl-2',3',4',5',6'-d5
CAS 1219798-92-1
Molekylær formel C12 D5 Cl5
Mængde 0.005 g
Synonym 1,1'-Biphenyl-2,3,4,5,6-d5, 2',3',4',5',6'-pentachloro- (ACI), 2',3',4',5',6'-Pentachloro-1,1'-biphenyl-2,3,4,5,6-d5 (ACI)
InChI formel InChI=1 S/C12H5Cl5/c13-8-7(6-4-2-1-3-5-6)9(14)11(16)12(17)10(8)15/h1-5 H/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2c(Cl)c(Cl)c(Cl)c(Cl)c2Cl
IUPAC navn 1,2,3,4,5-pentachloro-6-(2,3,4,5,6-pentadeuteriophenyl)benzene
Molekylvægt (g/mol) 331.4639
Formel vægt 328.9148 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity