Få mere at vide
2-[3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one Dihydrochloride, Mikromol™
Beskrivelse
2-[3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-[3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one Dihydrochloride |
| CAS | 2413877-03-7 |
| Analyt- eller komponentnavne | 2-[3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H22 Cl N5 O . 2 Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2-[3-[4-(4-Chlorophenyl)piperazin-1-yl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one Dihydrochloride, Trazodone Hydrochloride Imp. C (BP) as Dihydrochloride, Trazodone Related Compound C as Dihydrochloride, Trazodone Hydrochloride Impurity C, Trazodone Impurity C |
| InChI formel | InChI=1S/C19H22ClN5O.2ClH/c20-16-5-7-17(8-6-16)23-14-12-22(13-15-23)9-3-11-25-19(26)24-10-2-1-4-18(24)21-25;;/h1-2,4-8,10H,3,9,11-15H2;2*1H |
| SMIL | Cl.Cl.Clc1ccc(cc1)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2 |
| IUPAC navn | 2-[3-[4-(4-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;dihydrochloride |
| Vis mere |