Få mere at vide
2,3,4,4',5-Pentachlorobiphenyl-2',3',5',6'-d4, TRC
Artikelnummer. 30410699
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410699 5 mg
30422316 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410699 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2382015MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1219799-24-2
Molekylær formel C12 2H4 H Cl5
Molekylvægt (g/mol) 330.46
Synonym 2,3,4,4',5-Pentachlorobiphenyl-2',3',5',6'-D4, 2',3',5',6'-D4-2,3,4,4',5-PeCB, PCB 114-d4, 2,3,4,4',5-Pentachloro-1,1'-biphenyl l-2',3',5',6'-d4, 2,3,4,4',5-Pentachlorobiphenyl D4, 2,3,4,5,4'-Pentachlorobiphenyl D4, 4'-Chloro-2,3,4,5-tetrachloro-2',3',5',6'-tetradeuteriobiphenyl, CB 114-d4, PCB 114 D4, 2,3,4,4',5-PentaCB--d4
IUPAC navn 1-chloro-2,3,5,6-tetradeuterio-4-(2,3,4,5-tetrachlorophenyl)benzene
SMIL [2H]c1c([2H])c(c([2H])c([2H])c1Cl)c2cc(Cl)c(Cl)c(Cl)c2Cl
Tekniske data
Kemisk navn eller materiale 2,3,4,4',5-Pentachlorobiphenyl-2',3',5',6'-D4
Mængde 5 mg
InChI formel InChI=1S/C12H5Cl5/c13-7-3-1-6(2-4-7)8-5-9(14)11(16)12(17)10(8)15/h1-5H/i1D,2D,3D,4D
Formel vægt 327.9085