Få mere at vide
2-3-[4-(3-Bromophenyl)piperazin-1-yl]propyl-1,2,4-triazolo[4,3-a]-pyridin-3(2H)-one Hydrochloride, Mikromol™
Artikelnummer. 30050623
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30050623 50 mg
1 options
Artikelnummer. 30050623 Leverandør Mikromol Leverandørnr. MM0421.04
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-3-[4-(3-Bromophenyl)piperazin-1-yl]propyl-1,2,4-triazolo[4,3-a]-pyridin-3(2H)-one Hydrochloride
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-3-[4-(3-Bromophenyl)piperazin-1-yl]propyl-1,2,4-triazolo[4,3-a]-pyridin-3(2H)-one Hydrochloride
CAS 1263278-80-3
Analyt- eller komponentnavne 2-3-[4-(3-Bromophenyl)piperazin-1-yl]propyl-1,2,4-triazolo[4,3-a]-pyridin-3(2H)-one Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C19 H22 Br N5 O . Cl H
Certificeringer/compliance ISO 17025
Synonym 2-3-[4-(3-Bromophenyl)piperazin-1-yl]propyl-1,2,4-triazolo[4,3-a]-pyridin-3(2H)-one Hydrochloride, Trazodone Hydrochloride Imp. D (BP) as Hydrochloride, Trazodone Related Compound D as Hydrochloride, Trazodone Hydrochloride Impurity D, Trazodone Impurity D
InChI formel InChI=1S/C19H22BrN5O.ClH/c20-16-5-3-6-17(15-16)23-13-11-22(12-14-23)8-4-10-25-19(26)24-9-2-1-7-18(24)21-25;/h1-3,5-7,9,15H,4,8,10-14H2;1H
SMIL Cl.Brc1cccc(c1)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2
IUPAC navn 2-[3-[4-(3-bromophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride
Molekylvægt (g/mol) 452.78
Mængde 50 mg
Formel vægt 451.0775 g/mol