missing translation for 'onlineSavingsMsg'
Learn More

2,3,3-Trimethylindolenine, 98%, Thermo Scientific Chemicals

Catalog Number p-206356
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1640-39-7
C11H13N
159.23
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine, 2,3,3-trimethyl-3h-indole, 3h-indole, 2,3,3-trimethyl, ccris 6607, 2,3,3-trimethyl-indolenin, 2,3,3-trimethyl-3-hydroindole, pubchem7445, rarechem ah bs 0130, 2,3-trimethylindolenine, 2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C
This item is not returnable. View return policy
1640-39-7
C11H13N
159.23
MFCD00005724
FLHJIAFUWHPJRT-UHFFFAOYSA-N
2,3,3-trimethylindolenine, 2,3,3-trimethyl-3h-indole, 3h-indole, 2,3,3-trimethyl, ccris 6607, 2,3,3-trimethyl-indolenin, 2,3,3-trimethyl-3-hydroindole, pubchem7445, rarechem ah bs 0130, 2,3-trimethylindolenine, 2,3,3-trimethyl-indole
15427
2,3,3-trimethylindole
CC1=NC2=CC=CC=C2C1(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Density 0.9900g/mL
Boiling Point 228°C to 229°C (744.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.545
Quantity 25 g
Beilstein 20, 324
Specific Gravity 0.99
Formula Weight 159.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 3, 3-Trimethylindolenine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,3-Trimethylindolenine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'