Få mere at vide
2,3,3',4,4',5-Hexachlorobiphenyl-2',6,6'-d3, TRC
Artikelnummer. 30404576
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30404576 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404576 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2909595MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,3',4,4',5-Hexachlorobiphenyl-2',6,6'-d3
CAS 1219798-44-3
Molekylær formel C12 D3 H Cl6
Mængde 5 mg
Synonym 1,1'-Biphenyl-2,2',6-d3, 3,3',4,4',5',6'-hexachloro-, 2,3,3',4,4',5-Hexachlorobiphenyl-2',6,6'-d3, PCB 156-d3, 2,3,3',4,4',5-Hexachlorobiphenyl-2',6,6'-D3
InChI formel InChI=1S/C12H4Cl6/c13-7-2-1-5(3-8(7)14)6-4-9(15)11(17)12(18)10(6)16/h1-4H/i1D,3D,4D
SMIL [2H]c1cc(Cl)c(Cl)c([2H])c1c2c([2H])c(Cl)c(Cl)c(Cl)c2Cl
IUPAC navn 1,2,3,4-tetrachloro-5-deuterio-6-(3,4-dichloro-2,6-dideuteriophenyl)benzene
Molekylvægt (g/mol) 363.9
Formel vægt 360.8632