Få mere at vide
2,3,3',4,4',5-Hexachlorobiphenyl-2',6,6'-d3, CDN
Artikelnummer. 30319868
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319868 0.01 g
30319185 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319868 Leverandør CDN Leverandørnr. CDND69140.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,3',4,4',5-Hexachlorobiphenyl-2',6,6'-d3
CAS 1219798-44-3
Molekylær formel C12 D3 H Cl6
Mængde 0.01 g
Synonym PCB No. 156 d3, 1,1'-Biphenyl-2,2',6-d3, 3,3',4,4',5',6'-hexachloro-, PCB 156-d3
InChI formel InChI=1 S/C12H4Cl6/c13-7-2-1-5(3-8(7)14)6-4-9(15)11(17)12(18)10(6)16/h1-4 H/i1D,3 D,4 D
SMIL [2 H]c1cc(Cl)c(Cl)c([2 H])c1c2c([2 H])c(Cl)c(Cl)c(Cl)c2Cl
IUPAC navn 1,2,3,4-tetrachloro-5-deuterio-6-(3,4-dichloro-2,6-dideuterio-phenyl)benzene
Molekylvægt (g/mol) 363.9
Formel vægt 360.8632 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity