Få mere at vide
2,3,3,4,4,5,5-d7-L-Proline, TRC
Artikelnummer. 30482520
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30482520 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482520 Leverandør Toronto Research Chemicals Leverandørnr. TRCL45565025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2,3,3,4,4,5,5-d7-L-Proline
CAS 39063-89-3
Molekylær formel C5H2D7NO2
Mængde 25 mg
InChI formel InChI=1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1/i1D2,2D2,3D2,4D
SMIL OC([C@]1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])N1)=O
Molekylvægt (g/mol) 122.17