missing translation for 'onlineSavingsMsg'
Learn More

2-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl] Sumatriptan (>90%), TRC

Product Code. 30134460
Change view
Click to view available options
Mængde:
2.5 mg
25 mg
100 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
30134460 100 mg -
17924735 2.5 mg 2.5mg
17972545 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134460 Supplier Toronto Research Chemicals Supplier No. TRCD446670100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545338-89-4
Molekylær formel C27 H37 N5 O2 S
Molekylvægt (g/mol) 495.68
Synonym 1H-Indole-5-methanesulfonamide, 3-[2-(dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-N-methyl-, [3-[2-(Dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide, Sumatriptan Succinate Imp. A (EP)
IUPAC navn 1-[3-[2-(dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide
SMIL CNS(=O)(=O)Cc1ccc2[nH]c(Cc3ccc4[nH]cc(CCN(C)C)c4c3)c(CCN(C)C)c2c1

Specifications

Kemisk navn eller materiale 2-[(3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl)methyl] Sumatriptan
Mængde 100 mg
InChI formel InChI=1S/C27H37N5O2S/c1-28-35(33,34)18-20-7-9-26-24(15-20)22(11-13-32(4)5)27(30-26)16-19-6-8-25-23(14-19)21(17-29-25)10-12-31(2)3/h6-9,14-15,17,28-30H,10-13,16,18H2,1-5H3
Formel vægt 495.2668
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.