Få mere at vide
[2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]phenyl]methanol, Mikromol™
Artikelnummer. 30051111
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051111 100 mg
30052116 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051111 Leverandør Mikromol Leverandørnr. MM0147.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
[2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]phenyl]methanol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]phenyl)methanol
CAS 57321-32-1
Analyt- eller komponentnavne (2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]phenyl)methanol
Forsendelsestilstand Room Temperature
Molekylær formel C18 H22 N2 O
Certificeringer/compliance ISO 17025
Synonym Mianserin Hydrochloride Imp. A (EP), Mianserin Imp. A (EP), [2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]phenyl]methanol, Mianserin Hydrochloride Impurity A, Mianserin Impurity A
InChI formel InChI=1S/C18H22N2O/c1-19-11-12-20(17-10-6-5-9-16(17)14-21)18(13-19)15-7-3-2-4-8-15/h2-10,18,21H,11-14H2,1H3
SMIL OCC1=CC=CC=C1N2C(C3=CC=CC=C3)CN(C)CC2
IUPAC navn [2-(4-methyl-2-phenylpiperazin-1-yl)phenyl]methanol
Molekylvægt (g/mol) 282.38
Mængde 100 mg
Formel vægt 282.1732 g/mol