Få mere at vide
2-[(2R)-2-Hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-1H-isoindole-1,3(2H)-dione, Mikromol™
Artikelnummer. 30049850
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049850 100 mg
30051388 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049850 Leverandør Mikromol Leverandørnr. MM3629.09
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-[(2R)-2-Hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-1H-isoindole-1,3(2H)-dione
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-[(2R)-2-Hydroxy-3-[(4-(3-oxo-4-morpholinyl)phenyl)amino]propyl]-1H-isoindole-1,3(2H)-dione
CAS 446292-07-5
Analyt- eller komponentnavne 2-[(2R)-2-Hydroxy-3-[(4-(3-oxo-4-morpholinyl)phenyl)amino]propyl]-1H-isoindole-1,3(2H)-dione
Forsendelsestilstand Room Temperature
Molekylær formel C21 H21 N3 O5
Certificeringer/compliance ISO 9001
Synonym 2-[(2R)-2-Hydroxy-3-[[4-(3-oxo-4-morpholinyl)phenyl]amino]propyl]-1H-isoindole-1,3(2H)-dione
InChI formel InChI=1S/C21H21N3O5/c25-16(12-24-20(27)17-3-1-2-4-18(17)21(24)28)11-22-14-5-7-15(8-6-14)23-9-10-29-13-19(23)26/h1-8,16,22,25H,9-13H2/t16-/m1/s1
SMIL O[C@H](CNc1ccc(cc1)N2CCOCC2=O)CN3C(=O)c4ccccc4C3=O
IUPAC navn 2-[(2R)-2-hydroxy-3-[4-(3-oxomorpholin-4-yl)anilino]propyl]isoindole-1,3-dione
Molekylvægt (g/mol) 395.41
Mængde 100 mg
Formel vægt 395.1481 g/mol