missing translation for 'onlineSavingsMsg'
Få mere at vide

2,2'-Thiobis(ethanol)-13C4, TRC

Artikelnummer. 30425259
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425259 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30425259 Leverandør Toronto Research Chemicals Leverandørnr. TRCT26484210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2,2'-Thiobis(ethanol)-13C4
CAS 138847-50-4
Molekylær formel 13C4H10O2S
Mængde 10 mg
Synonym 2,2'-Thiobis(ethan-1-ol-1,2-13C2)
InChI formel InChI=1S/C4H10O2S/c5-1-3-7-4-2-6/h5-6H,1-4H2/i1+1,2+1,3+1,4+1
SMIL O[13CH2][13CH2]S[13CH2][13CH2]O
IUPAC navn 2-(2-hydroxy(1,2-^{13}C_{2})ethylsulfanyl)(1,2-^{13}C_{2})ethanol
Molekylvægt (g/mol) 126.15
Formel vægt 126.0536
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.