Få mere at vide
2-(2-t-Boc-aminoethoxy)anisole-d3, TRC
Artikelnummer. 30348695
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30348695 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348695 Leverandør Toronto Research Chemicals Leverandørnr. TRCB620902500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(2-t-Boc-aminoethoxy)anisole-d3
CAS 1219179-77-7
Molekylær formel C14H18D3NO4
Mængde 500 mg
Synonym 2-(3-Methoxyphenoxy)ethan-t-boc-amine-d3
InChI formel InChI=1S/C14H21NO4/c1-14(2,3)19-13(16)15-9-10-18-12-8-6-5-7-11(12)17-4/h5-8H,9-10H2,1-4H3,(H,15,16)/i4D3
SMIL [2H]C([2H])([2H])Oc1ccccc1OCCNC(=O)OC(C)(C)C
IUPAC navn tert-butyl N-[2-[2-(trideuteriomethoxy)phenoxy]ethyl]carbamate
Molekylvægt (g/mol) 270.34
Formel vægt 270.1659